O1-tert-butyl O2-methyl (2S)-4-(trifluoromethyl)-2,5-dihydropyrrole-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPBV0643-1G
Supplier: Pharmablock (USA)
O1-tert-butyl O2-methyl (2S)-4-(trifluoromethyl)-2,5-dihydropyrrole-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPBV0643-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
1,5-Diaminopentane 98%
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone 99.5% for analysis
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Isobutanol ≥99% ACS
Supplier: Spectrum Chemical
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals
n-Hexane
Supplier: Roth Carl
Dichloromethane 99.9% stabilised, AcroSeal® for biochemistry
Supplier: Thermo Fisher Scientific
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals