6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine-2,4-diol 2,2,2-trifluoroacetic acid 1 * 10 g
Catalog # PHBLPBV0199-2-10G
Supplier: Pharmablock (USA)
6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine-2,4-diol 2,2,2-trifluoroacetic acid 1 * 10 g
Catalog # PHBLPBV0199-2-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals