benzyl cis-4,4-dioxo-1,2,3,4a,5,7,8,8a-octahydropyrido[3,4-b][1,4]thiazine-6-carboxylate hydrochloride 1 * 1 g
Catalog # PHBLPBV0093-1-1G
Supplier: Pharmablock (USA)
benzyl cis-4,4-dioxo-1,2,3,4a,5,7,8,8a-octahydropyrido[3,4-b][1,4]thiazine-6-carboxylate hydrochloride 1 * 1 g
Catalog # PHBLPBV0093-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99%
Supplier: KLINIPATH
Carbon disulphide ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 2,5 L
Supplier: PanReac AppliChem
Pyridine, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Butyl acetate 99+%, extra pure
Supplier: Thermo Fisher Scientific