5-(difluoromethyl)-1-methyl-pyrazol-3-amine hydrochloride 1 * 1 g
PHBLPBUD073-1-1G
5-(difluoromethyl)-1-methyl-pyrazol-3-amine hydrochloride 1 * 1 g
PHBLPBUD073-1-1G
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals