O5-tert-butyl O2-ethyl rel-(2S,3aS,6aS)-2,3,3a,4,6,6a-hexahydro-1H-pyrrolo[3,4-b]pyrrole-2,5-dicarboxylate 1 * 1 g
Catalog # PHBLPBUC197-1G
Supplier: Pharmablock (USA)
O5-tert-butyl O2-ethyl rel-(2S,3aS,6aS)-2,3,3a,4,6,6a-hexahydro-1H-pyrrolo[3,4-b]pyrrole-2,5-dicarboxylate 1 * 1 g
Catalog # PHBLPBUC197-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals