methyl 4-amino-3-[[(2S)-oxetan-2-yl]methylamino]benzoate 1 * 1 g
Catalog # PHBLPBU7783-1G
Supplier: Pharmablock (USA)
methyl 4-amino-3-[[(2S)-oxetan-2-yl]methylamino]benzoate 1 * 1 g
Catalog # PHBLPBU7783-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
1-Butylboronic acid, Supelco®
Supplier: Merck
Petroleum spirit, 100…120 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) 95+% for spectroscopy
Supplier: Thermo Fisher Scientific
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals