methyl 2-[(1R,3R)-1-hydroxy-4-methyl-3-(methylamino)pentyl]thiazole-4-carboxylate 1 * 1 g
Catalog # PHBLPBU7009-1G
Supplier: Pharmablock (USA)
methyl 2-[(1R,3R)-1-hydroxy-4-methyl-3-(methylamino)pentyl]thiazole-4-carboxylate 1 * 1 g
Catalog # PHBLPBU7009-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
2-Propanol ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Ethylene glycol, high purity
Supplier: VWR Chemicals
2-Methylbutane for HPLC
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1,10-Phenanthroline monohydrate, Supelco®
Supplier: Merck
2-Methylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals