3-[[(3R)-4-(difluoromethyl)-2,2-difluoro-3-hydroxy-1,1-dioxo-3H-benzothiophen-5-yl]oxy]-5-fluoro-benzonitrile 1 * 1 g
Catalog # PHBLPBU5451-1G
Supplier: Pharmablock (USA)
3-[[(3R)-4-(difluoromethyl)-2,2-difluoro-3-hydroxy-1,1-dioxo-3H-benzothiophen-5-yl]oxy]-5-fluoro-benzonitrile 1 * 1 g
Catalog # PHBLPBU5451-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Hydrogen peroxide 35%, J.T.Baker®
Supplier: Avantor
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals