1-chloro-4-fluoro-5-methoxy-2-(trifluoromethoxy)benzene 1 * 1 g
Catalog # PHBLPBU3469-1G
Supplier: Pharmablock (USA)
1-chloro-4-fluoro-5-methoxy-2-(trifluoromethoxy)benzene 1 * 1 g
Catalog # PHBLPBU3469-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
N-HEPTANE FOR LIQUID CHROMATOGRAPHY 1 * 185 L
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulphoxide, EMPLURA®, Supelco®
Supplier: Merck
Cyclohexane
Supplier: Roth Carl
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Reag. USP, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND