1-chloro-3-fluoro-5-methoxy-2-(trifluoromethoxy)benzene 1 * 1 g
Catalog # PHBLPBU3236-1G
Supplier: Pharmablock (USA)
1-chloro-3-fluoro-5-methoxy-2-(trifluoromethoxy)benzene 1 * 1 g
Catalog # PHBLPBU3236-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Ethanolamine, EMSURE® for analysis, Supelco®
Supplier: Merck
n-Hexane ≥99%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
n-Hexane ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Butyl acetate ≥99.5% (by GC), EMSURE® for analysis, Supelco®
Supplier: Merck
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Acetonitrile, extra dry 99.9+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Turpentine oil
Supplier: Roth Carl
n-Hexane
Supplier: Roth Carl
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Water, TraceSELECT™ for trace analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Diethyl ether stabilised, BAKER, J.T.Baker®
Supplier: Avantor
Ethyl methanesulphonate 99%
Supplier: Thermo Fisher Scientific
Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific