1-[(2R,3R,4R)-4-[tert-butyl(dimethyl)silyl]oxy-3-fluoro-5,5-bis(hydroxymethyl)tetrahydrofuran-2-yl]pyrimidine-2,4-dione 1 * 1 g
Catalog # PHBLPBU2402-1G
Supplier: Pharmablock (USA)
1-[(2R,3R,4R)-4-[tert-butyl(dimethyl)silyl]oxy-3-fluoro-5,5-bis(hydroxymethyl)tetrahydrofuran-2-yl]pyrimidine-2,4-dione 1 * 1 g
Catalog # PHBLPBU2402-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran, EMPLURA®
Supplier: Merck
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Diethyl ether stabilised, TECHNICAL, rectified
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific