methyl (2R,3R)-2-methyl-1-[(1R)-1-phenylethyl]pyrrolidine-3-carboxylate 1 * 1 g
PHBLPBU1752-1G
methyl (2R,3R)-2-methyl-1-[(1R)-1-phenylethyl]pyrrolidine-3-carboxylate 1 * 1 g
PHBLPBU1752-1G
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals