(1R,2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)cyclopentanecarboxylic acid 1 * 1 g
Catalog # PHBLPBU1658-1G
Supplier: Pharmablock (USA)
(1R,2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)cyclopentanecarboxylic acid 1 * 1 g
Catalog # PHBLPBU1658-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Formamide, EMSURE® for analysis, Supelco®
Supplier: Merck
Methanol, EMPLURA®, Supelco®
Supplier: Merck
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Acetone ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
n-Pentane ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane ≥99.0%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanolamine 99%
Supplier: Thermo Fisher Scientific
Sodium diethyldithiocarbamate trihydrate, Supelco®
Supplier: Merck
Toluene, extra dry 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific