(2-{4,4-dimethyl-9-oxo-1,10-diazatricyclo[6.4.0.0,]dodeca-2(6),7-dien-10-yl}-4-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2- yl)phenyl)methyl acetate 1 * 1 g
Catalog # PHBLPBU1085-1G
Supplier: Pharmablock (USA)
(2-{4,4-dimethyl-9-oxo-1,10-diazatricyclo[6.4.0.0,]dodeca-2(6),7-dien-10-yl}-4-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2- yl)phenyl)methyl acetate 1 * 1 g
Catalog # PHBLPBU1085-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Toluene ≥98%, TECHNICAL
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals