methyl trans-4-(methylamino)cyclohexanecarboxylate hydrochloride 1 * 1 g
Catalog # PHBLPBU0504-1-1G
Leverancier: Pharmablock (USA)
methyl trans-4-(methylamino)cyclohexanecarboxylate hydrochloride 1 * 1 g
Catalog # PHBLPBU0504-1-1G
Leverancier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Leverancier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Leverancier: Merck
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Leverancier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Leverancier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Leverancier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Leverancier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM®, super gradient grade for HPLC, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals