O8-tert-butyl O2-methyl 8-azabicyclo[3.2.1]oct-2-ene-2,8-dicarboxylate 1 * 1 g
PHBLPBU0310-1G
O8-tert-butyl O2-methyl 8-azabicyclo[3.2.1]oct-2-ene-2,8-dicarboxylate 1 * 1 g
PHBLPBU0310-1G
Frequently Bought Together
4-Methyl-2-pentanone ≥97%, TECHNICAL
: VWR Chemicals
ACETONITRILE EMPLURA® 1 * 4 L
: Merck
1,2-Dichlorobenzene 99+% for HPLC
: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
: PanReac AppliChem
Toluene ≥99.3%, Laboratory Reagent
: Honeywell Chemicals