2-(bromomethyl)-1,3-dimethylbenzene 1 * 5 g
PHBLPBTYC1629-5G
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile 99.9%, ECD tested for residue analysis
: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
Formamide 99.5+%, extra pure
: Thermo Fisher Scientific
Cyclohexane 99.8% for HPLC
: Thermo Fisher Scientific
1,2-Dichloroethane 99+%, pure
: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
: PanReac AppliChem