[(2R,4aR,6S,7R,8S,8aR)-7-benzoyloxy-2-phenyl-6-(p-tolylsulfanyl)-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-8-yl] benzoate 1 * 1 g
Catalog # PHBLPBTS020-1G
Supplier: Pharmablock (USA)
[(2R,4aR,6S,7R,8S,8aR)-7-benzoyloxy-2-phenyl-6-(p-tolylsulfanyl)-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-8-yl] benzoate 1 * 1 g
Catalog # PHBLPBTS020-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
4-Aminobenzoic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Cyclohexane ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Cyclohexane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 2,5 L
Supplier: PanReac AppliChem
Diisopropyl ether 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Acetonitrile Reag. Ph. Eur., gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals
N,N-DIMETHYLFORMAMIDE AR | 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Dichloromethane 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific