2,2,2-trifluoro-1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone 1 * 1 g
Catalog # PHBLPBTD465-1G
Supplier: Pharmablock (USA)
2,2,2-trifluoro-1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone 1 * 1 g
Catalog # PHBLPBTD465-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
n-Hexane, EMPLURA®, Supelco®
Supplier: Merck
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Butyl acetate ≥99.5%, Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
N-Methyl-2-pyrrolidone (NMP) ≥99%
Supplier: Thermo Fisher Scientific
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Ethylene glycol ≥99% (by GC), pure
Supplier: PanReac AppliChem
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific