dicyclohexyl-[2-(2,6-dimethoxyphenyl)phenyl]phosphane (N-methyl-2-phenyl-anilino)-methylsulfonyloxy-palladium 1 * 1 g
Catalog # PHBLPBTA266-1G
Supplier: Pharmablock (USA)
dicyclohexyl-[2-(2,6-dimethoxyphenyl)phenyl]phosphane (N-methyl-2-phenyl-anilino)-methylsulfonyloxy-palladium 1 * 1 g
Catalog # PHBLPBTA266-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanolamine ≥99%, GPR RECTAPUR® for liquid scintillation counting
Supplier: VWR Chemicals
4-Methyl-2-pentanone ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane 99+%, ECD tested for pesticide analysis for residue analysis
Supplier: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific