(2S)-2-[(2S,3R,4R)-3,4-dihydroxy-5-oxo-tetrahydrofuran-2-yl]-2-hydroxy-acetic acid hydrate 1 * 1 g
Catalog # PHBLPBT8034-9-1G
Supplier: Pharmablock (USA)
(2S)-2-[(2S,3R,4R)-3,4-dihydroxy-5-oxo-tetrahydrofuran-2-yl]-2-hydroxy-acetic acid hydrate 1 * 1 g
Catalog # PHBLPBT8034-9-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Butanol NF
Supplier: Spectrum Chemical
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
1,2-Dichlorobenzene 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Diethyl ether stabilised, BAKER, J.T.Baker®
Supplier: Avantor
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals