tert-butyl (6R)-3-bromo-6-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-5-carboxylate 1 * 1 g
Catalog # PHBLPBSQA035-1G
Supplier: Pharmablock (USA)
tert-butyl (6R)-3-bromo-6-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-5-carboxylate 1 * 1 g
Catalog # PHBLPBSQA035-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Xylene - Mixture of isomers technical grade 1 * 5 L
Supplier: PanReac AppliChem
Isobutyronitrile 99%
Supplier: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals