tert-butyl N-methyl-N-[2-(methylamino)ethyl]carbamate 1 * 10 g
Catalog # PHBLPBS65219-10G
Supplier: Pharmablock (USA)
tert-butyl N-methyl-N-[2-(methylamino)ethyl]carbamate 1 * 10 g
Catalog # PHBLPBS65219-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-(Trimethylsilyl)imidazole, Supelco®
Supplier: Merck
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem