N-[9-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenyl-methoxy]methyl]-4-[2-cyanoethoxy-(diisopropylamino)phosphanyl]oxy-3-fluoro-tetra hydrofuran-2-yl]purin-6-yl]benzamide 1 * 1 g
Catalog # PHBLPBQS677-1G
Supplier: Pharmablock (USA)
N-[9-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenyl-methoxy]methyl]-4-[2-cyanoethoxy-(diisopropylamino)phosphanyl]oxy-3-fluoro-tetra hydrofuran-2-yl]purin-6-yl]benzamide 1 * 1 g
Catalog # PHBLPBQS677-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Hexane anhydrous, 95 % 1 * 250 mL
Supplier: Merck
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Methanol ACS, gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Hydrindantin dihydrate, Supelco®
Supplier: Merck
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Propanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific