tert-butyl rel-(3aR,5r,6aS)-5-amino-3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrole-2-carboxylate hydrochloride 1 * 1 g
Catalog # PHBLN2012216201-1G
Supplier: Pharmablock (USA)
tert-butyl rel-(3aR,5r,6aS)-5-amino-3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrole-2-carboxylate hydrochloride 1 * 1 g
Catalog # PHBLN2012216201-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
N,N-Dimethylformamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Chloroform, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Isooctane for UV, IR, HPLC, ACS 1 * 1 L
Supplier: PanReac AppliChem
N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran, anhydrous 99.9% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Petroleum spirit, 35…60 °C ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals