O1-tert-butyl O2-methyl (2R)-4,4-difluoropyrrolidine-1,2-dicarboxylate 1 * 1 g
PHBLPBN20121531-1G
O1-tert-butyl O2-methyl (2R)-4,4-difluoropyrrolidine-1,2-dicarboxylate 1 * 1 g
PHBLPBN20121531-1G
Frequently Bought Together
Water, TECHNICAL, demineralised
: VWR Chemicals
Dimethyl Sulfoxide EMPROVE EXPERT Ph Eur,USP 1 * 1 L
: MERCK PRODUCTION CHEMICALS
Cyclohexane 99+% for spectroscopy
: Thermo Fisher Scientific
Toluene ≥99.5% ACS
: Thermo Fisher Scientific
Triethylamine for HPLC
: Thermo Fisher Scientific
1,5-Diaminopentane 98%
: Thermo Fisher Scientific
Diethyl ether ≥99% stabilised for HPLC
: Thermo Fisher Scientific
Acetonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
: Honeywell Chemicals
Diethyl Ether stabilized with ethanol for UV, IR, HPLC 1 * 1.000 mL
: PanReac AppliChem
n-Pentane 98%, Reagent Grade
: Honeywell Chemicals