tert-butyl trans-3-methoxy-4-(methylamino)pyrrolidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBN20121071-1G
Supplier: Pharmablock (USA)
tert-butyl trans-3-methoxy-4-(methylamino)pyrrolidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBN20121071-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Hydrogen peroxide 35%, J.T.Baker®
Supplier: Avantor
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.0% USP, Ph. Eur.
Supplier: VWR Chemicals
1,3-Propanediol ≥99.7%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Acetone ≥99.8%, SupraSolv® for GC-MS, Supelco®
Supplier: Merck