(2S,4S)-1-[(tert-butoxy)carbonyl]-4-(trifluoromethyl)pyrrolidine-2-carboxylic acid 1 * 1 g
Catalog # PHBLPBN20120696-1G
Supplier: Pharmablock (USA)
(2S,4S)-1-[(tert-butoxy)carbonyl]-4-(trifluoromethyl)pyrrolidine-2-carboxylic acid 1 * 1 g
Catalog # PHBLPBN20120696-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1,6-Dibromohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
1-Butanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
4-Methyl-2-pentanone ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
FAM Benzine DIN 51635, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
Chloroform ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific