3-bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine 1 * 10 g
Catalog # PHBLPBN2011972-10G
Supplier: Pharmablock (USA)
3-bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine 1 * 10 g
Catalog # PHBLPBN2011972-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99%
Supplier: Thermo Fisher Scientific
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
tert-Butanol, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Fisher Scientific