(2R,3R)-2,3-dihydroxybutanedioic acid ethyl (3R)-piperidine-3-carboxylate 1 * 50 g
PHBLN20118060250G
(2R,3R)-2,3-dihydroxybutanedioic acid ethyl (3R)-piperidine-3-carboxylate 1 * 50 g
PHBLN20118060250G
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals