tert-butyl rel-(3aS,5s,6aR)-5-(methylamino)-3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrole-2-carboxylate 1 * 1 g
Catalog # PHBLPBMY0004-1G
Supplier: Pharmablock (USA)
tert-butyl rel-(3aS,5s,6aR)-5-(methylamino)-3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrole-2-carboxylate 1 * 1 g
Catalog # PHBLPBMY0004-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
(±)-2-Butanol ≥99% analytical reagent
Supplier: VWR Chemicals
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Methanol, SupraSolv® for GC-MS
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Pentane ≥95%, TECHNICAL
Supplier: VWR Chemicals
tert-Butyl methyl ether, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
Trichloroethylene ≥98%
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
2-Butoxyethanol ≥99%
Supplier: Thermo Fisher Scientific