4-(1H-Pyrazol-4-yl)-7-((2-(trimethylsilyl)ethoxy)-methyl)-7H-pyrrolo[2,3-d]pyrimidine 1 * 1 g
Catalog # PHBLPBLJ7323-1G
Supplier: Pharmablock (USA)
4-(1H-Pyrazol-4-yl)-7-((2-(trimethylsilyl)ethoxy)-methyl)-7H-pyrrolo[2,3-d]pyrimidine 1 * 1 g
Catalog # PHBLPBLJ7323-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-MS, Supelco®
Supplier: Merck
(±)-2-Butanol, EMSURE® for analysis, Supelco®
Supplier: Merck
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals