6-bromo-2-chloro-8-cyclopentyl-5-methyl-7H,8H-pyrido[2,3-d]pyrimidin-7-one 1 * 10 g
Catalog # PHBLPBLJ7236-10G
Supplier: Pharmablock (USA)
6-bromo-2-chloro-8-cyclopentyl-5-methyl-7H,8H-pyrido[2,3-d]pyrimidin-7-one 1 * 10 g
Catalog # PHBLPBLJ7236-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
Carbon disulphide ≥99.9%, Reagent Grade low in benzene
Supplier: Honeywell Chemicals
Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
n-Pentane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.0% USP, Ph. Eur.
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.5% (by GC), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Pyridine, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Toluene ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Acetonitrile, Prepsolv® for preparative chromatography
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific