5-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1h-pyrrolo[2,3-b]pyridine 1 * 1 g
Catalog # PHBLPBLJ3346-1G
Supplier: Pharmablock (USA)
5-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1h-pyrrolo[2,3-b]pyridine 1 * 1 g
Catalog # PHBLPBLJ3346-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Diethylene glycol monomethyl ether, Sigma-Aldrich®
Supplier: Merck
N-Ethyldiisopropylamine ≥99.5% for peptide synthesis
Supplier: VWR Chemicals
Hydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
1-Propanol, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene ≥99.5% ACS
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.7% for gas chromatography
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetone ≥99%
Supplier: Thermo Fisher Scientific
Isobutyronitrile 99%
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers), extra pure
Supplier: Thermo Fisher Scientific
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific