tert-butyl 2-(8-bromo-7-methyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-1-yl)acetate 1 * 1 g
Catalog # PHBLPBLHA032-1G
Supplier: Pharmablock (USA)
tert-butyl 2-(8-bromo-7-methyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-1-yl)acetate 1 * 1 g
Catalog # PHBLPBLHA032-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
Isobutyronitrile 99%
Supplier: Thermo Fisher Scientific
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Supplier: Honeywell Chemicals
Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Propanol ACS
Supplier: Thermo Fisher Scientific