benzyl 2-[[(2S)-2-[[(2R)-2-[[2-(tert-butoxycarbonylamino)acetyl]amino]-3-phenyl-propanoyl]amino]-4-methyl-pentanoyl]amino]acetate 1 * 1 g
Catalog # PHBLPBJL8033-1G
Supplier: Pharmablock (USA)
benzyl 2-[[(2S)-2-[[(2R)-2-[[2-(tert-butoxycarbonylamino)acetyl]amino]-3-phenyl-propanoyl]amino]-4-methyl-pentanoyl]amino]acetate 1 * 1 g
Catalog # PHBLPBJL8033-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol 90% 90% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Ethanol ≥96% (v/v)
Supplier: KLINIPATH
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals