(6R)-6-(4-bromophenyl)-4-[(1S)-1-phenylethyl]morpholin-3-one 1 * 1 g
Catalog # PHBLPBJL661-1G
Supplier: Pharmablock (USA)
(6R)-6-(4-bromophenyl)-4-[(1S)-1-phenylethyl]morpholin-3-one 1 * 1 g
Catalog # PHBLPBJL661-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethylene glycol, EMPLURA®, Supelco®
Supplier: Merck
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Cyclohexane, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
(±)-2-Butanol ≥99% analytical reagent
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
tert-Butanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Methanol, SupraSolv® for GC-MS
Supplier: Merck
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Acetonitrile with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Glycerine, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific