(3aS,6R,6aS)-2,2,6-trimethyl-3a,5,6,6a-tetrahydrocyclopenta[d][1,3]dioxol-4-one 1 * 1 g
Catalog # PHBLPBJL152-1G
Supplier: Pharmablock (USA)
(3aS,6R,6aS)-2,2,6-trimethyl-3a,5,6,6a-tetrahydrocyclopenta[d][1,3]dioxol-4-one 1 * 1 g
Catalog # PHBLPBJL152-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetone 99.9% (by GC, corrected for water content), ECD tested halocarbons free grade for residue analysis
Supplier: Thermo Fisher Scientific
Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 1 L
Supplier: PanReac AppliChem
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific