1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole 1 * 1 g
Catalog # PHBLPBJL1165-1G
Leverancier: Pharmablock (USA)
1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole 1 * 1 g
Catalog # PHBLPBJL1165-1G
Leverancier: Pharmablock (USA)
Frequently Bought Together
1-Butanol, EMPROVE® ESSENTIAL NF, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Acetone ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Leverancier: VWR Chemicals
Cyclohexane, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Leverancier: Merck
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
4-Aminobenzoic acid for synthesis, Sigma-Aldrich®
Leverancier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Leverancier: VWR Chemicals
Methanol ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Leverancier: VWR Chemicals
Butyl acetate ≥99.5% for HPLC
Leverancier: Thermo Fisher Scientific
N-HEPTANE 95% CHROMAR HPLC | 2,5 DM3 1 * 2,5 L
Leverancier: MACRON AVANTOR BRAND
Methanol Reag. Ph. Eur., ACS, ISO, Burdick & Jackson™
Leverancier: Honeywell Chemicals
Ethyl acetate, anhydrous (max. 0.005% H₂O) ≥99.8%
Leverancier: VWR Chemicals
n-Heptane 95%
Leverancier: Thermo Fisher Scientific
9-Fluorenylmethyl chloroformate (Fmoc chloride), Supelco®
Leverancier: Merck
n-Hexane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Leverancier: Avantor
Dichloromethane, anhydrous ≥99.7% stabilised
Leverancier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Leverancier: Honeywell Chemicals