methyl trans-4-[(tert-butoxycarbonylamino)methyl]cyclohexanecarboxylate 1 * 5 g
Catalog # PHBLPBJL100287-5G
Supplier: Pharmablock (USA)
methyl trans-4-[(tert-butoxycarbonylamino)methyl]cyclohexanecarboxylate 1 * 5 g
Catalog # PHBLPBJL100287-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
4-Methyl-2-pentanone 99+% for HPLC
Supplier: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Pyridine, dried ≥99.5% (by GC), Puriss. Reag. Ph. Eur. (≤0.0075% H₂O), Riedel-de Haën™
Supplier: Honeywell Chemicals
Methyl ethyl ketone ACS
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
2-Butoxyethanol ≥99%
Supplier: Thermo Fisher Scientific