tert-butyl 4-[4-(methylamino)pyrazol-1-yl]piperidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBHD274-1G
Supplier: Pharmablock (USA)
tert-butyl 4-[4-(methylamino)pyrazol-1-yl]piperidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBHD274-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Acetone ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
1-Propanol ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, Ultrapure for HPLC
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Hexanol 98%, Reagent Grade
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Diethyl ether ≥99.5% (by GC) stabilised, Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene 99.5% ACS for spectroscopy
Supplier: Thermo Fisher Scientific