1-(5-bromo-4-hexyl-2-thienyl)-5-methyl-2,8-dioxa-5-azonia-1-boranuidabicyclo[3.3.0]octane-3,7-dione 1 * 1 g
Catalog # PHBLPBHA623-1G
Supplier: Pharmablock (USA)
1-(5-bromo-4-hexyl-2-thienyl)-5-methyl-2,8-dioxa-5-azonia-1-boranuidabicyclo[3.3.0]octane-3,7-dione 1 * 1 g
Catalog # PHBLPBHA623-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Acetone, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Chloroform, LiChrosolv® for HPLC
Supplier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
1,2-Dichloroethane ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
n-Hexane, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals