N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]acetamide 1 * 1 g
Catalog # PHBLPBGJ1410-1G
Supplier: Pharmablock (USA)
N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]acetamide 1 * 1 g
Catalog # PHBLPBGJ1410-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Acetonitrile ≥99.7%, Environmental Grade
Supplier: Thermo Fisher Scientific
1-Butylboronic acid, Supelco®
Supplier: Merck
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dichloromethane 99.9% stabilised, AcroSeal® for biochemistry
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals