tert-butyl 3-(7-chloro-8-fluoro-5-methoxy-2-methylsulfanyl-pyrido[4,3-d]pyrimidin-4-yl)-3,8-diazabicyclo[3.2.1]octane-8-carboxylate 1 * 1 g
Catalog # PHBLPBG0584-1G
Supplier: Pharmablock (USA)
tert-butyl 3-(7-chloro-8-fluoro-5-methoxy-2-methylsulfanyl-pyrido[4,3-d]pyrimidin-4-yl)-3,8-diazabicyclo[3.2.1]octane-8-carboxylate 1 * 1 g
Catalog # PHBLPBG0584-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Hexane ≥95% for spectrophotometry
Supplier: Thermo Fisher Scientific
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised for HPLC
Supplier: Thermo Fisher Scientific