2-chloro-5-nitrobenzene-1-sulphonyl chloride 1 * 5 g
Catalog # PHBLPBES0055-5G
Supplier: Pharmablock (USA)
2-chloro-5-nitrobenzene-1-sulphonyl chloride 1 * 5 g
Catalog # PHBLPBES0055-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals