2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 1 * 1 g
PHBLPBEB10650-1G
2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 1 * 1 g
PHBLPBEB10650-1G
Frequently Bought Together
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
: PanReac AppliChem
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabiliser free, AcroSeal®
: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Methanol ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
: VWR Chemicals
1-Hexanol 98%, Reagent Grade
: Honeywell Chemicals
Petroleum spirit, 80…110 °C
: Roth Carl
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
: Thermo Fisher Scientific
(±)-1,2-Propanediol ACS
: Thermo Fisher Scientific
n-Pentane, anhydrous 99+%, extra pure
: Thermo Fisher Scientific
tert-Butanol 99.5%, extra pure
: Thermo Fisher Scientific
Xylene - Mixture of isomeres (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
: PanReac AppliChem