tert-butyl 5-bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate 1 * 1 g
Catalog # PHBLPBEB10430-1G
Supplier: Pharmablock (USA)
tert-butyl 5-bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate 1 * 1 g
Catalog # PHBLPBEB10430-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Cyclohexane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
(±)-2-Butanol ≥99% analytical reagent
Supplier: VWR Chemicals
Toluene ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Toluene, anhydrous (max. 0.005% H₂O) ≥99.8%, AnalaR NORMAPUR® for synthesis
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific