N-cyclohexyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine 1 * 1 g
Catalog # PHBLPBEB0700-1G
Supplier: Pharmablock (USA)
N-cyclohexyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine 1 * 1 g
Catalog # PHBLPBEB0700-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., VOLUSOL® titration solvent
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Dimethyl sulphoxide, EMPLURA®, Supelco®
Supplier: Merck
Toluene ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals