(1S,4S)-1-METHYL-7-(TRIFLUOROMETHYL)-3,4 1 * 1 g
Catalog # PHBLPBDL895-1-1G
Supplier: Pharmablock (USA)
(1S,4S)-1-METHYL-7-(TRIFLUOROMETHYL)-3,4 1 * 1 g
Catalog # PHBLPBDL895-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
METHANOL ANHYDROUS, 99.8 % 1 * 1 L
Supplier: Merck
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
[EN]DIBUTYL ETHER ANHYDROUS, 99.3 % 1 * 2 L
Supplier: Merck
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
N,N-Dimethylaceetamide 99%, pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.9% stabilised, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Tolueen, GPR RECTAPUR® low in sulphur
Supplier: VWR Chemicals
n-Heptaan ∼ 99% (n-heptane) for HPLC
Supplier: Thermo Fisher Scientific
Diethylether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Hexane (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific
Cyclohexaan ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Aceton ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
2-Propanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor