[(2R,3S,4R,5R,6R)-3,4-diacetoxy-6-bromo-5-[(2,2,2-trifluoroacetyl)amino]tetrahydropyran-2-yl]methyl acetate 1 * 1 g
Catalog # PHBLPBDL887-1G
Supplier: Pharmablock (USA)
[(2R,3S,4R,5R,6R)-3,4-diacetoxy-6-bromo-5-[(2,2,2-trifluoroacetyl)amino]tetrahydropyran-2-yl]methyl acetate 1 * 1 g
Catalog # PHBLPBDL887-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals